Ekspektoratbeholdere, Nunc™
Leverandør: Thermo Fisher Scientific
549263
391-7798EA 2100 NOK
391-7798
Ekspektoratbeholdere, Nunc™
Prøvecontainere Beholdere, generelt bruk
Polystyrol.
  • Low profile container can be shipped by normal mail
  • Large opening facilitates secure sample placement
  • Lowered centre of lid facilitates scraping off samples when the lid is removed
  • Ribs on lid and container for easy grip
For sampling and transport of diagnostic specimens.
Sertifiseringer: Containers comply with 95 kPa test described in UN packaging instructions 602/650 for transport of infectious substances and diagnostic specimens.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904.