Cell culture flasks with standard or filter caps, CELLSTAR®
Leverandør: Greiner Bio-One
658170_120 690160_200 660175_50 690170_200 658175_120 661175_40 661160_40 660160_50 690175_200
391-3106EA 2950 NOK
391-3106 391-3103 391-3148 391-3104 391-3146 392-0013 392-0012 391-3109 391-3143
Cell culture flasks with standard or filter caps, CELLSTAR®
Kolber Celledyrkningsflasker
PS, sterile.
  • Improved cell adhesion due to physical surface treatment
  • Easy-open filter cap guarantees optimal gas exchange whilst minimising the risk of contamination
  • Stacking ridges on the top of the flask makes them easy to stack and ensures that they stand firmly in the incubator
  • Graduated on both sides for easier filling
  • Choice of flat or high profile design of the 175 cm² flask, allows for variability of media volume
These standard cell culture flasks are supplied with either standard screw caps or capillary pore membrane filter caps (poreMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat ISO 14001 (241002); ISO 50001 (241002)