Cell culture flasks for automated systems, CELLSTAR® AutoFlask™
Leverandør: Greiner Bio-One
779160_100 779190_100
392-0017EA 11925 NOK
392-0017 392-0088
Cell culture flasks for automated systems, CELLSTAR® AutoFlask™
Kolber Celledyrkningsflasker
PS, sterile.
  • Compatible with a wide range of cell culture and liquid handling systems
  • Pre-scored multiple entry septum assures sterility of flask contents throughout processing
  • Centrifugation pocket enables separation of cells from supernatant in the flask
  • Integrated hydrophobic filter facilitates gas exchange during cell cultivation
  • Colour coding allows an easy identification of cell culture AutoFlask (red) and suspension culture AutoFlask (white)
These flasks are specifically designed for automated cell culture. For the cultivation of adherentMore Product Information