Microplates, streptavidin coated, 96- and 384-well
Leverandør: Greiner Bio-One
655990_40 781990_40 781997_40 655997_40 781995_40
392-0031EA 29525 NOK
392-0031 737-0082 737-0084 392-0034 738-0174
Microplates, streptavidin coated, 96- and 384-well
Mikroplater Immunologi-mikroplater og strips
PS, without lid. Streptavidin coated solid phases serve as reliable binding surfaces for all types of biotinylated molecules. Numerous ligands can be biotinylated simply and reliably, and due to the low molecular weight of biotin (244 Da), the functionality of the molecules is normally not impaired. Thus streptavidin-coated solid phases make it possible to simply and rapidly isolate, determine and quantify components from a reaction mixture. By immobilising the biotinylated substance, it is also possible to reproduce complete reaction chains on a streptavidin solid phase, for example, enzymeMore Product Information