Cell Culture Flasks, CELLCOAT®
Leverandør: Greiner Bio-One
690920_10 658940_50 658950_50 661940_40 661950_40 690940_50 690950_50
392-0257EA 4300 NOK
392-0257 392-0273 392-0274 392-0275 392-0276 392-0280 392-0281
Cell Culture Flasks, CELLCOAT®
Kolber Celledyrkningsflasker
Clear PS flasks with CELLCOAT® surface treatment and red filter screw caps. Coated with proteins of the extracellular matrix (Collagen Type I or Fibronectin) or synthetic proteins (Poly-DLysine).
  • Made of high-grade polystyrene
  • Free of detectable DNases, RNases, human DNA, endotoxins; non-cytotoxic
  • Canted neck for optimal access
  • Printed graduation on both sides for easier filling
  • Improved adhesion and cell proliferation
  • Reduced-serum or serum-free cultivation
  • Improved growth of primary cells
The specific design facilitates an optimal access of theMore Product Information