Celledyrkningsplate, 96-brønner, Corning®
Leverandør: Corning
3879
392-0291EA 2625 NOK
392-0291
Celledyrkningsplate, 96-brønner, Corning®
Mikroplater Celledyrkningsplater
PS, klare, med lokk.
  • Corning® CellBIND® surface increases wetability for more even and consistent cell attachment
  • Ultra-Low Attachment plates feature a covalently bound hydrogel layer that minimises cell attachment, protein absorption and cellular activation
  • Sterilised by gamma irradiation
  • Certified non-pyrogenic
Recommended working volume: 75 to 200 µl.