VWR®, Cell culture media, Ham's F12
Leverandør: VWR Chemicals
392-0418EA 140 NOK
392-0418
VWR®, Cell culture media, Ham's F12
Media, Cellekultur
Ham's F12 was developed for growth of primary rat hepatocytes and rat prostate epithelial cells. Also used in a clonal toxicity assay using CHO cells. It is frequently used with supplements for serum-free cultures to support the growth of cells of rodent origin, or as a cloning medium for the culture of myeloma and hybrid cells.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Media Base Ham's F12 Nutrient Mixture
Kalsium Yes
HEPES No
L-Glutamine Yes
Magnesium Yes
Phenol red Yes
Sodium bicarbonate Yes
Sodium pyruvate Yes
Stable glutamine No