VWR®, Cell culture media, McCoy's 5A medium
Leverandør: VWR Chemicals
392-0420EA 305 NOK
392-0420
VWR®, Cell culture media, McCoy's 5A medium
Media, Cellekultur
McCoy’s media were originally formulated for growth and support of lymphocytes. This final modification produced a medium identical to RPMI 1629. McCoy's 5A media supports the indefinite proliferation of Walker 256 carcinoma cells. In addition, it is excellent for the propagation of leukocytes, biopsy tissues, a broad range of human and rat normal or transformed cell types, the most current primary and continuous cell lines.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Media Base McCoy's 5A Medium
Kalsium Yes
HEPES No
L-Glutamine Yes
Magnesium Yes
Phenol red Yes
Sodium bicarbonate Yes
Sodium pyruvate No
Stable glutamine No