VWR®, Phosphate Buffered Saline (PBS), pH 7,4
Leverandør: VWR Chemicals
392-0442EA 115 NOK
392-0442
VWR®, Phosphate Buffered Saline (PBS), pH 7,4
Buffere Buffere, cellekultur
Phosphate Buffered Saline (PBS) is a buffer solution containing sodium chloride, sodium phosphate and potassium chloride. PBS helps to maintain a constant pH, is isotonic and non toxic to cells, and provide cells with water and essential inorganic ions. This formulation does not contain calcium and magnesium.
  • PBS is used in a variety of laboratory applications:
  • Washing cells - prior to DNA/RNA extraction
  • Reagent preparation/dilution
  • Transporting cells
  • Diluent of human biological samples
Note: Please refer to the technical data sheet for recommended use.