Natriumtiosulfat pentahydrat 99.5-101.0% (by iodometry), krystaller, BAKER ANALYZED® ACS, J.T.Baker®

Leverandør: Avantor

3946-01EA 750 NOK
3946-01 3946-07
Natriumtiosulfat pentahydrat 99.5-101.0% (by iodometry), krystaller, BAKER ANALYZED® ACS, J.T.Baker®
Natriumtiosulfat pentahydrat
Formel: Na₂S₂O₃.5H₂O
Smeltepunkt: 48 °C
Tetthet: 1,67 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149186
CAS-nummer: 10102-17-7
EINECS: 231-867-5
Merck Indeks: 12,08844

Bestill nå

Spesifikasjon Testresultater

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use 99.5 - 101.0 %
Assay (Na₂S₂O₃·5H₂O) (by iodometry) 99.5 - 101.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 6.0 - 8.4
Nitrogen Compounds (as N) ≤0.002 %
Sulfate and Sulfite (as SO₄) ≤0.1 %
Sulfide (S) Passes Test
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR