Titanium(IV) oxide sulphate sulphuric acid hydrate

Leverandør: Thermo Fisher Scientific
Danger

39591.36 39591.A3
39591.36EA 1550 NOK
39591.36 39591.A3
Titanium(IV) oxide sulphate sulphuric acid hydrate
Titanium(IV) oxide sulphate sulphuric acid hydrate

CAS No.: 13825-74-6 Molecular Formula: TiOSO4.xH2O + H2SO4 Physical Form: -10 Mesh Powder UN No.: UN3260 MDL No.: MFCD00011270

Formel: TiOSO₄·xH₂O+H₂SO₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011270
CAS-nummer: 13825-74-6
EINECS: 237-523-0
ADR: 8,II
Merck Indeks: 14,9475

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR