Gold(I) sodium thiosulphate hydrate ≥99.9% (metals basis)

Leverandør: Thermo Fisher Scientific

39741.03 39741.06 39741.14
39741.03EA 2825 NOK
39741.03 39741.06 39741.14
Gold(I) sodium thiosulphate hydrate ≥99.9% (metals basis)
Gold(I) sodium thiosulphate hydrate

Purity: 99.9% CAS No.: 15283-45-1 Molecular Formula: Na3Au(S2O3)2.xH2O Formula Weight: 490.19(anhy) Physical Form: Powder Density: 3.09 Appearance: White Melting Point: dec. Solubility: Soluble in water. Insoluble in ethanol and most other organic solvents MDL No.: MFCD00046176

Formel: AuH₂Na₃O₆S₄·xH₂O
Smeltepunkt: Dec.
Tetthet: 3,09
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00046176
CAS-nummer: 15283-45-1
EINECS: 239-324-4
Merck Indeks: 14,4519

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR