EDTA dinatrium salt standardised solution 0.0575 M (0.115N) vandig løsning

Leverandør: Thermo Fisher Scientific

39782.K2 39782.K7
39782.K2EA 635 NOK
39782.K2 39782.K7
EDTA dinatrium salt standardised solution 0.0575 M (0.115N) vandig løsning
EDTA dinatrium salt

CAS No.: 139-33-3 Molecular Formula: (NaO2CCH2)2NCH2CH2N(CH2CO2H)2 Formula Weight: 336.22 Physical Form: Liquid Odour: Odourless MDL No.: MFCD00070672

Formel: C₁₀H₁₄N₂Na₂O₈
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00070672
CAS-nummer: 139-33-3
EINECS: 205-358-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR