Karbondisulfid ≥99.9% ACS

Leverandør: Thermo Scientific
Danger

39785.AY
39785.AYEA 3175 NOK
39785.AY
Karbondisulfid ≥99.9% ACS
Karbondisulfid
Formel: CS₂
MW: 76,14 g/mol
Kokepunkt: 46 °C
Smeltepunkt: –112 °C
Tetthet: 1,263
Flash punkt: –30 °C (–22 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011321
CAS-nummer: 75-15-0
EINECS: 200-843-6
FN: 1131
ADR: 3,I
Merck Indeks: 14,1811

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR