Tinn(II)klorid dihydrat 98-103%, krystaller, BAKER ANALYZED® ACS, J.T.Baker®

Leverandør: Avantor
Danger

3980-01EA 3950 NOK
3980-01 3980-05
Tinn(II)klorid dihydrat 98-103%, krystaller, BAKER ANALYZED® ACS, J.T.Baker®
Tinn(II)klorid dihydrat

Stannous chloride, dihydrate, crystal suitable for mercury determination.

Formel: SnCl₂·2H₂O
Kokepunkt: 623 °C (1013 hPa)
Smeltepunkt: 246 °C
Tetthet: 2,71 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149863
CAS-nummer: 10025-69-1
EINECS: 231-868-0
FN: 3260
ADR: 8,III
Merck Indeks: 13,08861

Bestill nå

Spesifikasjon Testresultater

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use 98.0 - 103.0%
Assay (SnCl₂·2H₂O) 98.0 - 103.0 %
Solubility in HCl Passes Test
Sulfate (SO₄) Passes Test
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤0.003 %
Lead (Pb) ≤0.01 %
Magnesium (Mg) ≤0.01 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.010 %
Trace Impurities - Arsenic (As) ≤2.000 ppm
Trace Impurities - Mercury (Hg) ≤0.050 ppm
Meets Reagent Specifications for testing USP/NF monographs

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR