Palladium 5% on strontium carbonate, pulver, reduced

Leverandør: Thermo Fisher Scientific

39819.04 39819.09 39819.18
39819.04EA 810 NOK
39819.04 39819.09 39819.18
Palladium 5% on strontium carbonate, pulver, reduced
Palladium
Formel: Pd
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00192595
CAS-nummer: 7440-05-3

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR