Trifluoreddiksyre 0,05% v/v aqueous solution, LiChrosolv® HPLC, Supelco®

Leverandør: Merck

Synonymer: TFA

MQ100

4.80170.2500
4.80170.2500EA 1060 NOK
4.80170.2500
Trifluoreddiksyre 0,05% v/v aqueous solution, LiChrosolv® HPLC, Supelco®
Trifluoreddiksyre
Formel: C₂HF₃O₂
MW: 114.02 g/mol
Kokepunkt: 71.8
Smeltepunkt: -15
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004169
CAS-nummer: 76-05-1
EINECS: 200-929-3
Merck Indeks: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR