1-Ethyl-3-methylimidazolium chloride, Sigma-Aldrich®

Leverandør: Merck
Warning

Synonymer: 1-Ethyl-3-methyl-1H-imidazol-3-ium chloride

M-Clarity™ quality level = MQ200

4.90054.1000
4.90054.1000EA 0 NOK
4.90054.1000
1-Ethyl-3-methylimidazolium chloride, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium chloride
Formel: C₆H₁₁ClN₂
Smeltepunkt: ∼80 °C
Tetthet: ∼1,1 g/cm³ (20 °C)
Flash punkt: 186 °C
MDL nummer: MFCD00074843
CAS-nummer: 65039-09-0

Bestill nå

Spesifikasjon Testresultater

Melting Point 77 - 79 °C
Molar Mass 146.62 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR