PMA (Propidium monoazide) DNA/RNA binding dye
Leverandør: Biotium
Synonymer: 3-Amino-8-azido-5-{3-[diethyl(methyl)ammonio]propyl}-6-phenylphenanthridinium, 3-Amino-5-[3-(methyldiethylaminio)propyl]-6-phenyl-8-azidophenanthridinium, Q16821519, SCHEMBL17399073
40013
40013.EA 3150 NOK
40013.
PMA (Propidium monoazide) DNA/RNA binding dye
PMA (Propidium monoazide)
PMA (propidium monoazide) is a high-affinity photoreactive DNA binding dye invented by scientists at Biotium for viability PCR of bacteria and other organisms. The dye is weakly fluorescent by itself but becomes more fluorescent after binding to nucleic acids.
  • Abs = 464 nm (before photolysis)
  • Abs/Em = 510/610 nm (following photolysis and covalent attachment to DNA/RNA)
  • Dark red solid soluble in water
  • Store at 4°C and protect from light at all times
It preferentially binds to dsDNA with high affinity. Upon photolysis, the dye covalently reacts with DNA, thus resulting in permanent DNA modification. The dye is cell membrane-impermeable, and thus can be selectively used to modify only exposed DNA from dead cells while leaving DNA from viable cells intact. This feature makes the dye highly useful in the selective detection of viable pathogenic cells by quantitative real-time PCR in the presence dead cells whose DNA has been PMA-modified and thus can not be amplified.
Since Biotium first developed PMA dye, there have been numerous publications on the use of the dye in pathogenic bacterial detection related to food and water safety, medical diagnosis and biodefense. Check the Downloads section to view the PMA&PMaxx™ Reference List and the PMA&PMAxx™ Validated Bacterial Strains List. PMA dye is also available in H2O (40019), a convenient ready-to-use format.
For photoactivation of PMA dye, we recommend our PMA-Lite™ LED Photolysis Device for microcentrifuge tubes, or the Glo-Plate Blue™ LED Illuminator for larger tubes or 96-well plates.