Biotin-11-uridine-5'-triphosphate, tetralithium salt (Biotin-11-UTP) 10 mM buffer (Tris-HCl, pH 7.5)
Leverandør: Biotium
40033
40033.EA 5075 NOK
40033.
Biotin-11-uridine-5'-triphosphate, tetralithium salt (Biotin-11-UTP) 10 mM buffer (Tris-HCl, pH 7.5)
Biotin-11-uridine-5'-triphosphate, tetralithium salt (Biotin-11-UTP)
Biotin-11-UTP (biotin-11-uridine-5'-triphosphate, tetralithium salt) can be enzymatically incorporated into RNA. The number 11 is the number of atoms in the linker between biotin and UTP.