Biotin-20-UTP 10 mM buffer (Tris-HCl, pH 7.5)
Leverandør: Biotium
Synonymer: Biotin-20-uridine-5'-triphosphate, tetralithium salt
40034
40034.EA 6125 NOK
40034.
Biotin-20-UTP 10 mM buffer (Tris-HCl, pH 7.5)
Biotin-20-UTP
Biotin-20-UTP can be enzymatically incorporated into RNA. The number 20 is the number of atoms in the linker between biotin and UTP.