Biotin-11-CTP (Biotin-11-cytidine-5′-triphosphate, tetralithium salt) 10 mM buffer (TRIS-HCl pH 7.5)
Leverandør: Biotium
40035
40035.EA 4350 NOK
40035.
Biotin-11-CTP (Biotin-11-cytidine-5′-triphosphate, tetralithium salt) 10 mM buffer (TRIS-HCl pH 7.5)
Biotin-11-CTP (Biotin-11-cytidine-5′-triphosphate, tetralithium salt)
Biotin-11-CTP (Biotin-11-cytidine-5′-triphosphate, tetralithium salt) can be enzymatically incorporated into RNA. The number 11 is the number of atoms in the linker between biotin and CTP.