Biotin-11-2′-deoxycytidine-5′-triphosphate, tetralithium salt (Biotin-11-dCTP) 1 mM TE Buffer (pH 7.5)

Leverandør: Biotium

40036
40036.EA 6075 NOK
40036.
Biotin-11-2′-deoxycytidine-5′-triphosphate, tetralithium salt (Biotin-11-dCTP) 1 mM TE Buffer (pH 7.5)
Biotin-11-2′-deoxycytidine-5′-triphosphate, tetralithium salt (Biotin-11-dCTP)

Biotin-11-dCTP (Biotin-11-2′-deoxycytidine-5′-triphosphate, tetralithium salt) can be enzymatically incorporated into DNA via nick translation, random priming, or 3′-end terminal labeling. The number 11 is the number of atoms in the linker between biotin and dCTP.

Formel: C₂₈H₄₂N₇O₁₆P₃SLi₄
MW: 885.42 g/mol
Lagringstemperatur: Fryseskap
CAS-nummer: 136632-30-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR