Acridine orange (chlorid salt) 10 mg/ml water, high_purity (AO)

Leverandør: Biotium
Danger

Synonymer: AO, C.I. 46005, 3,6-Bis(dimethylamino)acridine hydrochloride, Basic Orange 14, N,N,N’,N’-Tetramethyl-3,6-acridinediamine hydrochloride, 3,6-Acridinediamine, Acridine orange hydrochloride

40039
40039.EA 1325 NOK
40039.
Acridine orange (chlorid salt) 10 mg/ml water, high_purity (AO)
Acridine orange (chlorid salt)

Acridine orange (AO) stains dsDNA green (525 nm) and RNA or single stranded DNA red (650 nm). The dye is membrane-permeant and its nucleic acid binding property has been used for cell-cycle studies. Acridine orange has also been used for the detection of microorganisms in cerebrospinal fluid and other clinical specimens. We offer a highly purified form of acridine orange while most of the other commercially available grades of AO are either in zinc chloride complex form or of low purity.


  • λEx/λEm = 500/526 nm (DNA), 460/650 nm (RNA)
  • Yellow solution
  • Store at 4°C and protect from light

Formel: C₁₇H₁₉N₃·HCl·xH₂O
MW: 301.81 g/mol (anhy)
MDL nummer: MFCD00012660
CAS-nummer: 65-61-2
EINECS: 233-353-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40dc52c9a2ccf35ab469f6f3af7273bff3f3d2e3&vid=b6324af55fb9fc3b94fdd1a60ac80649fc3cdfb9&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR