dNTP set
Leverandør: Biotium
40052
40052.EA 3075 NOK
40052.
dNTP set
Nukleinsyre reagenser Nukleotider
Highly purified dNTPs (>99% by HPLC) in purified water. Each deoxynucleotide solution (dATP, dCTP, dGTP, dTTP) is provided in a separate vial at 100 mm.
The dNTPs are suitable for use in PCR, cDNA synthesis, sequencing, and other molecular biology applications.