Cyanine 647-dUTP, lyophilised blue solid
Leverandør: Biotium
40065
40065.EA 3800 NOK
40065.
Cyanine 647-dUTP, lyophilised blue solid
Cyanine 647-dUTP
Cyanine 647-dUTP can be used to synthesise labeled DNA probes for in-situ hybridization, microarray or blotting techniques.
Far-red fluorescent Cyanine 647 is structurally identical to Cy®5 and has absorption/emission at 650/665 nm.
NB: Store Cyanine 647-dUTP at –20 °C or –70 °C, protected from light.
Cy®5 dye is a registered trademark of GE Healthcare.