CF® Dye dCTP conjugates
Leverandør: Biotium
40028 40027 40058 40055 40056 40057 40066 40067 40068 40027-T 40028-T 40055-T 40056-T 40057-T 40058-T 40066-T 40067-T 40068-T
40028.EA 7600 NOK
40028. 40027. 40058. 40055. 40056. 40057. 40066. 40067. 40068. 40027-T 40028-T 40055-T 40056-T 40057-T 40058-T 40066-T 40067-T 40068-T
CF® Dye dCTP conjugates
Systemer for proteinkobling og -merking
CF® dye-dCTP can be used to synthesise probes for in situ hybridisation, microarray, or to synthesise labelled DNA probes for in situ hybridisation and nucleic acid blotting applications.
  • Synthesise exceptionally bright and photostable fluorescent DNA probes
  • Detect by fluorescence microscopy, flow cytometry, or in gel
  • Compatible with fixed cells or tissue sections
  • Choice of nine CF® dye colours from green to far-red
  • CF® dyes have superior brightness, photostability, and are water-soluble
NB: CF™594 dCTP is not compatible with PCR labelling of DNA probes.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Maks eksitasjon 555 nm 650 nm 562 nm 593 nm 527 nm 541 nm 642 nm 490 nm 663 nm
max emisjon 565 nm 665 nm 583 nm 614 nm 558 nm 560 nm 662 nm 515 nm 682 nm
Vert