Cadmium molybdenum oxide ≥99.5% (metals basis)

Leverandør: Thermo Scientific
Danger

40202.30
40202.30EA 8425 NOK
40202.30
Cadmium molybdenum oxide ≥99.5% (metals basis)
Cadmium molybdenum oxide

Purity: 99.5% CAS No.: 13972-68-4 Molecular Formula: CdMoO4 Formula Weight: 272.35 Physical Form: Powder Odour: Odourless Melting Point: ca 1250° Solubility: Slightly soluble in water. Soluble in acid UN No.: UN2570 MDL No.: MFCD00015997


Applications: Electronic and optical applications

Formel: CdMoO₄
MW: 272,35 g/mol
Smeltepunkt: ∼ 1250 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00015997
CAS-nummer: 13972-68-4
EINECS: 237-752-6
FN: 2570
ADR: 6.1,III

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR