Palladium ≥99.98% (metals basis), metalltråd, Ø 1.0 mm

Leverandør: Thermo Fisher Scientific

40212.BS 40212.BW 40212.CB
40212.BSEA 4875 NOK
40212.BS 40212.BW 40212.CB
Palladium ≥99.98% (metals basis), metalltråd, Ø 1.0 mm
Palladium

Purity: 99.98+% CAS No.: 7440-05-3 Physical Form: ≈9.44g/m Odour: Odourless MDL No.: MFCD00011167

Formel: Pd
Kokepunkt: 3167 °C (1013 hPa)
Smeltepunkt: 1555 °C
Tetthet: 12,02 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011167
CAS-nummer: 7440-05-3
EINECS: 231-115-6
Merck Indeks: 12,07121

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR