Saltsyre 32%

Leverandør: Honeywell Chemicals
Danger

Synonymer: Hydrochloric acid solution, Muriatic acid

40233-2.5L
40233-2.5LEA 1650 NOK
40233-2.5L
Saltsyre 32%
Saltsyre
Formel: HCl
MW: 36.46 g/mol
Smeltepunkt: –30 °C
Tetthet: 1.18 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
FN: 1789
ADR: 8 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR