ICP-MS tuning standard solution for 6020 CLP-M, Specpure®

Leverandør: Thermo Fisher Scientific

40482.AEEA 2475 NOK
40482.AE
ICP-MS tuning standard solution for 6020 CLP-M, Specpure®
Standarder Kationstandarder Multi-Element Standards

UN No.: UN3264
MDL No.: MFCD00240453

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
MDL No. MFCD00240453

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR