ICP-MS stock standard solution A for 200,8, Rev. 4.4, Specpure®

Leverandør: Thermo Fisher Scientific

40485.AEEA 5925 NOK
40485.AE
ICP-MS stock standard solution A for 200,8, Rev. 4.4, Specpure®
Standarder Kationstandarder Multi-Element Standards

UN No.: UN3264
MDL No.: MFCD00803326

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR