3,4-Dimetylheksan, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ100

40512-1ML-F
40512-1ML-FEA 580 NOK
40512-1ML-F
3,4-Dimetylheksan, Supelco®
3,4-Dimetylheksan
Formel: C₈H₁₈
MW: 114.23 g/mol
Kokepunkt: 118
Smeltepunkt: –64 °C
Tetthet: 0.7152 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00039927
CAS-nummer: 583-48-2
EINECS: 209-504-7
FN: 1262
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9458a814164d4be4fa6c8851d75888b78fe919f&vid=24a32ecbcbb199e7b3a8e908cc46d663f2dba68a&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR