Supelco® TraceCERT® Organic Reference Standard, Simazine

Leverandør: Merck

40662-100MG
40662-100MGEA 980 NOK
40662-100MG
Supelco® TraceCERT® Organic Reference Standard, Simazine
Standarder Organiske standarder

Simazine CRM may also be used as given below: Development and validation of an ultrasonic-assisted dispersive liquid-liquid microextraction (UA-DLLME) method for sample extraction followed by gas chromatography-mass spectrometry based determination of endocrine-disrupting herbicides in the river and seawater samples. Determination of triazine pesticide residues in samples of apples pears carrots potatoes tomatoes avocado cucumber spinach bananas and oranges by modified QuEChERS and solid-phase extraction (SPE) procedures followed by liquid chromatography-photodiode array (LC-DAD) analysis. Multi-residue analysis of 67 pesticides in olive oil olives and avocado samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS) to evaluate various clean-up sorbents. Quantitation of 20 pesticides in eel and shrimp samples by a scheduled multiple reaction monitoring (sMRM) method and QuEChERS with EDTA for sample extraction and purification. Analysis of 35 micropollutants including pesticides in water samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS) after their solid-phase extraction. Simultaneous quantification of 58 pharmaceuticals and personal care products (PPCPs) and pesticides in water samples by solvent-based de-emulsification dispersive liquid-liquid microextraction (SD-DLLME) combined with liquid chromatography-tandem mass spectrometry (LCMS/MS).


Sertifiseringer: The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.


NB: P202 - P264 - P270 - P273 - P301 + P312 - P308 + P313

Bestill nå

SPESIFIKASJONER

Synonym(s) 2,4-Bis(ethylamino)-6-chloro-1,3,5-triazine
Environmentally Preferable
M-Clarity quality level MQ300
CAS-nr 122-34-9
MDL No. MFCD00023174
Molekylvekt 201,66
Hardhetsklasse beskrivelse H302 - H351 - H410
Fraktforhold STD
Lagringsforhold 2...8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR