Aluminium ≥99.1% (metals basis), ultrathin foil, Thickness: 0.8 µm

Leverandør: Thermo Fisher Scientific

40707.6X 40707.YD 40707.6M
40707.6XEA 6900 NOK
40707.6X 40707.YD 40707.6M
Aluminium ≥99.1% (metals basis), ultrathin foil, Thickness: 0.8 µm
Aluminium

Purity: 99.1% CAS No.: 7429-90-5 Physical Form: Not supported Odour: Odourless MDL No.: MFCD00134029

Formel: Al
Kokepunkt: 2327 °C (1013 hPa)
Smeltepunkt: 660 °C
Tetthet: 2,7 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00134029
CAS-nummer: 7429-90-5
EINECS: 231-072-3
Merck Indeks: 13,00321

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR