D(+)-Sakkarose, crystals NF, Multi-Compendial, J.T.Baker®

Leverandør: Avantor
Warning

Synonymer: β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose

4074-01EA 1400 NOK
4074-01 4074-05 4074-07 4074-19 4074-20
D(+)-Sakkarose, crystals NF, Multi-Compendial, J.T.Baker®
D(+)-Sakkarose
Formel: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Smeltepunkt: 169…170 °C
Tetthet: 1.59 g/cm³
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006626
CAS-nummer: 57-50-1
EINECS: 200-334-9
Merck Indeks: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

GMP Manufactured Product Passes Test
Meets J.P. Chemical Specifications Passes Test
Meets B.P. Chemical Specifications ≤45
Meets E.P. Chemical Specifications ≤35 µS/cm
Meets N.F. Requirements 66.3 - 67.0 Degree
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical ≤10 ppm
NF - Identification Passes Test
NF - Appearance of Solution Passes Test
NF - Color Value ≤45
NF - Conductivity ≤35 µS/cm
NF - Optical Rotation (+) 66.3 - 67.0 Degree
NF - Reducing Sugars Passes Test
NF - Sulfite (SO2) ≤10 ppm
NF - Loss on Drying at 105 ºC ≤0.1 %
EP/BP - Identification A (EP/BP) Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Conductivity, uS cm-1 ≤35
EP/BP - Optical Rotation (+) 66.3 - 67.0 Degree
EP/BP - Colour Value ≤45
EP/BP - Reducing Sugars Passes Test
EP/BP - Sulfite (SO₂) ≤10 ppm
EP/BP - Loss on Drying at 105°C ≤0.1 %
JP - Identification Passes Test
JP - Optical Rotation (+) 66.3 - 67.0 Degree
JP - Clarity of Solution Passes Test
JP - Color Value ≤45
JP - Sulfite (SO₂) ≤10 ppm
JP - Reducing Sugars Passes Test
JP - Conductivity, uS cm-1 ≤35
JP - Loss on Drying at 105°C ≤0.1 %
Endotoxin Concentration (EU/g) ≤250
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.
Store in a tightly closed container.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR