Calbiochem® Human recombinant interleukin-1 beta
Leverandør: Merck Millipore Calbiochem
407615-5
407615-5EA 12525 NOK
407615-5
Calbiochem® Human recombinant interleukin-1 beta
Proteiner
Recombinant, human interleukin-1ß expressed in E. coli. Major form of interleukin-1 secreted by monocytes and macrophages. Induces nitric oxide synthase in pancreatic and smooth muscle cells and suppresses apoptosis in rat ovarian follicles. Involved in inflammatory and immune responses and promotes wound healing.