Phytochemical standard, Neohesperidin, analytical standard, Supelco®

Leverandør: Merck

40927-10MG
40927-10MGEA 3025 NOK
40927-10MG
Phytochemical standard, Neohesperidin, analytical standard, Supelco®
Standarder Organiske standarder

Flavanone glycoside with neuroprotective and antioxidant properties. Does not inhibit oral carcinogenesis in a rat model unlike other citrus flavanones, but does exert a protective effect against gastritis and gastric lesions.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea0f912732b4e2b27505735dbf08c0e7d077a71&vid=5ea0f912732b4e2b27505735dbf08c0e7d077a71&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Beskrivelse Neohesperidin
CAS-nr 13241-33-3
Formel C₂₈H₃₄O₁₅
Lagringstemp 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR