Tris-(hydroksymetyl)aminometan ≥99.0% (dried basis), BAKER ANALYZED®, Biochemical Reagent for liquid chromatography, J.T.Baker®

Leverandør: Avantor


Management of Change (MOC) category = N

4099-07EA 0 NOK
4099-07
Tris-(hydroksymetyl)aminometan ≥99.0% (dried basis), BAKER ANALYZED®, Biochemical Reagent for liquid chromatography, J.T.Baker®
Tris-(hydroksymetyl)aminometan
Formel: NH₂C(CH₂OH)₃
Kokepunkt: 357 °C (1013 hPa)
Smeltepunkt: 169…172 °C
Tetthet: 1,328 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004679
CAS-nummer: 77-86-1
EINECS: 201-064-4
Merck Indeks: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use ≥99.0%
Appearance Passes Test
Assay (C₄H₁₁NO₃) (dried basis) ≥99.0 %
Absorbance of a 40% Solution - 280 nm ≤0.200
Absorbance of a 40% Solution - 400 nm ≤0.020
Insoluble Matter ≤0.005 %
Water (H₂O)(by Karl Fischer titrn) ≤2 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR