Tris-(hydroksymetyl)aminometan 99.0-101.0% USP, Multi-Compendial, J.T.Baker®

Leverandør: Avantor
Warning

4102-01EA 2275 NOK
4102-01 4102-05 4102-07 4102-09 4102-F9 4102-F7 4102-F8
Tris-(hydroksymetyl)aminometan 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Tris-(hydroksymetyl)aminometan
Formel: NH₂C(CH₂OH)₃
Kokepunkt: 357 °C (1013 hPa)
Smeltepunkt: 169…172 °C
Tetthet: 1,328 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004679
CAS-nummer: 77-86-1
EINECS: 201-064-4
Merck Indeks: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Meets JPC (1997) Chemical Specifications 99.0 - 101.0%
Meets B.P. Chemical Specifications Passes Test
Meets E.P. Chemical Specifications Passes Test
Meets U.S.P Requirements Passes Test
GMP Manufactured Product 10.0 - 11.5
CAUTION: For Manufacturing, processing or repackaging ≤1.0%
Bulk Pharmaceutical Chemical ≤0.1%
USP - Assay (C₄H₁₁NO₃) 99.0 - 101.0 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C Passes Test
USP - pH (1 in 20) 10.0 - 11.5
USP - Loss on Drying ≤1.0 %
USP - Residue on Ignition ≤0.1 %
USP - Melting Range(Lower) 168 - 172 °C
USP - Melting Range(Upper) 168 - 172 °C
EP/BP - Assay (C₄H₁₁NO₃) (dried basis) 99.0 - 100.5 %
EP/BP - Identification B (melting point) 168 - 174 °C
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - pH 10.0 - 11.5
EP/BP - Related Substances ≤1.0 %
EP/BP - Chloride (Cl) ≤100 ppm
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Loss on Drying at 105°C ≤0.5 %
EP/BP - Ash (sulfated) ≤0.1 %
JPC 1997 - Assay (C₄H₁₁NO₃) (dried basis) ≥99.0 %
JPC 1997 - Identification A Passes Test
JPC 1997 - Identification B Passes Test
JPC 1997 - pH 10.3 - 10.7
JPC 1997 - Melting Point 168 - 172 °C
JP - Clarity and Color of Solution Passes Test
JPC 1997 - Heavy Metals (as Pb) ≤8 ppm
JPC 1997 - Arsenic (As) ≤1.6 ppm
JPC 1997 - Loss on Drying ≤0.5 %
JPC 1997 - Residue on Ignition ≤0.10 %
Assay (C₄H₁₁NO₃) (dried basis) ≥99.9 %
Absorbance of a 1M Solution - 260 nm ≤0.06
Absorbance of a 1M Solution - 280 nm ≤0.06
Absorbance of a 1M Solution - 400 nm ≤0.01
Endotoxin Concentration (EU/g) ≤2.5
Iron (Fe) ≤1 ppm
Lead (Pb) ≤1 ppm
Ultrapure Bioreagent Specifications
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Manufactured exclusively for Avantor Performance Materials, LLC., by BioSpectra, Inc., Stroudsburg, PA
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR