Tris-(hydroksymetyl)aminometan hydroklorid ≥99.0% Made from Tromethamine USP, Biotech Reagent, J.T.Baker®

Leverandør: Avantor
Warning

4106-01EA 4300 NOK
4106-01 4106-05 4106-07 4106-09 4106-F9 4106-F7
Tris-(hydroksymetyl)aminometan hydroklorid ≥99.0% Made from Tromethamine USP, Biotech Reagent, J.T.Baker®
Tris-(hydroksymetyl)aminometan hydroklorid

GMP manufactured product. The infrared spectrum conforms to the standard.

Formel: NH₂C(CH₂OH)₃·HCl
Smeltepunkt: 147…153 °C
Tetthet: 1,05 g/ml (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012590
CAS-nummer: 1185-53-1
EINECS: 214-684-5
Merck Indeks: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

GMP Manufactured Product ≥99.0%
CAUTION: For Manufacturing, processing or repackaging ≤0.06
Assay (NH₂C(CH₂OH)₃·HCl) ≥99.0 %
Appearance (colorless crystals to white crystalline powder) Passes Test
Absorbance of a 1M Solution - 260 nm ≤0.06
Absorbance of a 1M Solution - 280 nm ≤0.06
Absorbance of a 1M Solution - 400 nm ≤0.01
Identification (by IR) Passes Test
Loss on Drying at 105°C ≤0.5 %
Endotoxin Concentration (EU/g) ≤2.5
Melting Point 147 - 153 °C
pH of 0.5M Solution at 25°C 3.5 - 5.0
Trace Impurities - Arsenic (As) ≤10 ppm
Trace Impurities - Calcium (Ca) ≤10.0 ppm
Trace Impurities - Copper (Cu) ≤10 ppm
Trace Impurities - Iron (Fe) ≤10 ppm
Trace Impurities - Lead (Pb) ≤10 ppm
Trace Impurities - Magnesium (Mg) ≤10 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Manufactured exclusively for Avantor Performance Materials, LLC., by BioSpectra, Inc., Stroudsburg, PA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR