Periodic table mix 2 for ICP, 17 elements, TraceCERT®, Supelco®
Leverandør: Merck
41135-100ML
41135-100MLEA 2725 NOK
41135-100ML
Periodic table mix 2 for ICP, 17 elements, TraceCERT®, Supelco®
Standarder Kationstandarder Multi-Element Standards
10 mg/L: Au, Ge, Hf, Ir, Mo, Nb, Pd, Pt, Re, Rh, Ru, Sb, Sn, Ta, Ti, W, Zr in 5%hydrochloric acid and 1% hydrofluoric acid (contains HNO₃ traces)
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034