Mill, Tube Mill control
Leverandør: IKA
4180000 20000279 20000283
412-0203EA 32325 NOK
412-0203 412-0204 412-0205
Mill, Tube Mill control
Kverner
The Tube Mill control is a highly versatile small batch mill with disposable grinding chambers thereby eliminating cross-contamination and cleaning effort.
  • Easy and quiet operation with touch keypad to access various operating modes
  • Programmable and storable test conditions delivers reproducible test results
  • USB interface for control and documentation of all the parameters
  • Interval operation available 5 to 60 seconds
  • Stainless steel cutter/beater
  • Adjustable safety speed and time
  • Dust proof sealed housing
The grinding chamber and cover areMore Product Information