Supelco® TraceCERT® Organic Reference Standard, Tebutam
Leverandør: Merck
41397-100MG
41397-100MGEA 1275 NOK
41397-100MG
Supelco® TraceCERT® Organic Reference Standard, Tebutam
Standarder Organiske standarder
Tebutam CRM may also be used as given below:
Optimisation of sample extraction based on QuEChERS and dispersive-solid-phase extraction (d-SPE) clean-up for the multi-residue analysis of 120 pesticides 16 polycyclic aromatic hydrocarbons and 22 polychlorinated biphenyls by liquid and gas chromatography coupled with tandem mass spectrometry (MS/MS)
Evaluation of 250 pesticide residues extracted by QuEChERS method from 103 processed fruit samples using ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)
Determination of 90 pesticides 16 polycyclic aromatic hydrocarbons and 22 polychlorinated biphenyls in honey samples by using QuEChERS combined with dispersive solid-phase extraction (SPME) for sample pre-treatment and liquid chromatography-triple-quadrupole tandem mass spectrometry and gas chromatography-ion-trap tandem mass spectrometry for quantitation
Screening and quantification of 569 pesticides in honeysuckle samples by a combination of ultrahigh-performance liquid chromatography (UHPLC) and quadrupole-Orbitrap high-resolution mass spectrometry
Simultaneous determination of 134 pesticide residues in tea samples using d-SPE extraction with a multi-functional filter (MFF) followed by ultra-high performance liquid chromatography (UPLC) and quadrupole time-of-flight mass spectrometry (TOF-MS).
Sertifiseringer: The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.