Natrium 1000 mg/L Na i vandig (fra NaCl) Specpure® enkeltgrunnstoffsstandard til ionekromatografi

Leverandør: Thermo Fisher Scientific

41696.AEEA 1950 NOK
41696.AE
Natrium 1000 mg/L Na i vandig (fra NaCl) Specpure® enkeltgrunnstoffsstandard til ionekromatografi
Standarder Kationstandarder Sodium Standards

Molecular Formula: NaCl in H₂O
Physical Form: Liquid
MDL No.: MFCD00044631


Sertifiseringer: Traceable to NIST

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR