Nikkel ≥99% (metals basis), metalltråd, annealed, Ø 0.15 mm (0.006 in) (Nickel 200)

Leverandør: Thermo Fisher Scientific
Danger

41785.H7 41785.I1
41785.H7EA 1625 NOK
41785.H7 41785.I1
Nikkel ≥99% (metals basis), metalltråd, annealed, Ø 0.15 mm (0.006 in) (Nickel 200)
Nikkel

Form: ∼0.16g/m
Temper: soft

Formel: Ni
Kokepunkt: 3007 °C (1013 hPa)
Smeltepunkt: 1453 °C
Tetthet: 8,9 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011137
CAS-nummer: 7440-02-0
EINECS: 231-111-4
FN: 3089
ADR: 4.1,III
Merck Indeks: 12,06582

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR