Alizarin red S (sodium salt)

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: C.I. 58005

42040.06 42040.14
42040.06EA 605 NOK
42040.06 42040.14
Alizarin red S (sodium salt)
Alizarin red S (sodium salt)

CAS No.: 130-22-3 Molecular Formula: C14H7O7SNa Formula Weight: 342.27 Physical Form: Powder Appearance: Yellow to red Beilstein Reference: 4117090 Merck Reference: 14,8573 MDL No.: MFCD00013049

Formel: C₁₄H₇NaO₇S
MW: 342.26 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013049
CAS-nummer: 130-22-3
EINECS: 204-981-8
Merck Indeks: 14,8573

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR