Cresyl diphenyl phosphate (mixture of analogue) mixed isomers 94

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: Diphenyl p-tolyl phosphate, Diphenyl Cresyl Phosphate, Phosphoric Acid Cresyl Diphenyl Ester

42092.AE 42092.AP
42092.AEEA 630 NOK
42092.AE 42092.AP
Cresyl diphenyl phosphate (mixture of analogue) mixed isomers 94
Cresyl diphenyl phosphate (mixture of analogue)

Purity: 94% CAS No.: 26444-49-5 Molecular Formula: (CH3C6H4O)xP(O)(OC6H5)y Flash Point: 232°(450°F) Physical Form: Liquid Density: 1.21 Refractive Index: 1.5630 Appearance: Pale yellow Melting Point: -38° Boiling Point: 390° UN No.: UN3082 MDL No.: MFCD00014920

Formel: C₁₉H₁₇O₄P
MW: 340.3097 g/mol
Kokepunkt: 390 °C
Smeltepunkt: –38 °C
Tetthet: 1.21
Flash punkt: 232 °C (450 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00014920
CAS-nummer: 26444-49-5
EINECS: 247-693-8
FN: 3082
ADR: 9 III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR