Kobber ≥99.999% (metals basis), folie, Puratronic®, Thickness: 0.1 mm (0.004 in)

Leverandør: Thermo Fisher Scientific

42189.FI 42189.GH 42189.HG
42189.FIEA 1750 NOK
42189.FI 42189.GH 42189.HG
Kobber ≥99.999% (metals basis), folie, Puratronic®, Thickness: 0.1 mm (0.004 in)
Kobber

Purity: 99.999% CAS No.: 7440-50-8 Physical Form: ≈2.2g/50x50mm Odour: Odourless MDL No.: MFCD00010965

Formel: Cu
Kokepunkt: 2595 °C (1013 hPa)
Smeltepunkt: 1083 °C
Tetthet: 8,94 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010965
CAS-nummer: 7440-50-8
EINECS: 231-159-6
Merck Indeks: 13,02545

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR