VWR®, Bovine serum albumin (BSA)
Leverandør: VWR Chemicals
422361VEA 5200 NOK
422361V
VWR®, Bovine serum albumin (BSA)
Proteiner
A protease-free grade albumin for RIA and ELISA applications, produced by a heat-shock process in caprylic acid.
  • Protease-free
  • IgG-free
  • Moisture: 5%